missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse Wnt-5a Protein

Product Code. 18117580
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Unit Size:
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18117580 Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. 10µg
18385597 Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18117580 Supplier R&D Systems™ Supplier No. 645WN010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human/Mouse Wnt-5a Protein is derived from CHO. The Recombinant Human/Mouse Wnt-5a Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein.
Gene ID (Entrez) 7474
Molecular Weight (g/mol) 38 kDa
Name Wnt-5a
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived Wnt-5a protein Gln38-Lys380
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.