missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat UBE2M/Ubc12 Protein

Product Code. 18357477
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357477 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357477 Supplier R&D Systems™ Supplier No. E2656100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Recombinant Human UBE2M/Ubc12 is a member of the NEDD8-conjugating (E2) enzyme family that receives NEDD8 from a NEDD8-activating (E1) enzyme and subsequently interacts with a NEDD8 ligase (E3) to conjugate NEDD8 to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human UBE2M/Ubc12 concentration of 0.1-1 μM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P61081.1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, TCEP and Glycerol.
Gene ID (Entrez) 9040
Molecular Weight (g/mol) M.W. (theoretical): 21 kDa
Name UBE2M/Ubc12
Quantity 100 μg
Source E. coli-derived UBE2M/Ubc12 protein Met1 - Lys183
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol UBE2M
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein UBE2M/Ubc12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.