missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat Irisin Protein

Product Code. 18706767
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18706767

Brand: R&D Systems™ 8880IR025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human/Mouse/Rat Irisin Protein is derived from CHO. The Recombinant Human/Mouse/Rat Irisin Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NAU1
For Use With (Application) Bioactivity
Gene ID (Entrez) 252995
Molecular Weight (g/mol) Observed M.W.: 15-37kDa, reducing conditions, Theoretical M.W.: 13kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived Irisin/FNDC5 protein Asp32-Glu143,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FNDC5, FRCP2, FRCP2Fibronectin type III repeat-containing protein 2
Common Name Irisin/FNDC5
Gene Symbol FNDC5
Biological Activity Measured by its ability to induce p38 MAPK activation in 3T3-L1 mouse embryonic fibroblast adipose-like cells. 1μg/mL of Recombinant Human Irisin can effectively induce p38 MAPK activation.
Conjugate Unconjugated
Species Human, Mouse, Rat
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Asp32-Glu143, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.