missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat BMP-2 Protein

Product Code. p-4719153
Click to view available options
Quantity:
10 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid.
Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid.
Unit Size:
10µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18347747

Brand: R&D Systems™ 355BM010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 40-200 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid.
Gene ID (Entrez) 650
Molecular Weight (g/mol) 13 kDa (monomer)
Name BMP-2
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived BMP-2 protein Gln283-Arg396
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.