missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat BMP-2 Protein

Product Code. 18788076
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18788076 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18788076 Supplier R&D Systems™ Supplier No. 355BM100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 40-200 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12643
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid with BSA as
Gene ID (Entrez) 650
Molecular Weight (g/mol) 13 kDa (monomer)
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived BMP-2 protein Gln283-Arg396
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC
Gene Symbol BMP2
Conjugate Unconjugated
Cross Reactivity BMP-2
Species Human, Mouse, Rat
Reconstitution Reconstitute at 100-200 μg/mL in sterile 4 mM HCl containing at least 0.1% human or bovine serum albumin.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.