missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed)
Description
The Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed) is derived from E. coli. The Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed) has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
| Gene ID (Entrez) | 650 |
| Molecular Weight (g/mol) | 12.8 kDa (monomer) |
| Name | BMP-2 |
| Quantity | 10 μg |
| Source | E. coli-derived BMP-2 protein Ala284-Arg396 This form is not detected using human BMP-2 Quantikine ELISA (Catalog &Num; DBP200 ). |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?