missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human, Mouse IL-15 Fc, Mutant Protein [Biotin]

Product Code. 18372345 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372345 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372345 Supplier Novus Biologicals™ Supplier No. NBP318179

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration LYOPH
For Use With (Application) SDS-PAGE
Formulation Lyophilized from PBS
Gene ID (Entrez) 3600
Quantity 1 Vial
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.06 EU/μg protein (LAL test; Lonza).
Gene Alias IL-15MGC9721, interleukin 15, interleukin-15
Gene Symbol IL15
Research Category Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
Purity or Quality Grade >98%, by SDS-PAGE
Protein IL-15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.