missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human MMR/CD206 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Mannan is coated at 0.2 μg/mL (100 μL/well), the concentration of Recombinant Human MMR/CD206 that produces 50% of the optimal binding response is approximately 0.2-1.2 μg/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 4360 |
| Molecular Weight (g/mol) | 157 kDa |
| Name | MMR/CD206 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human MMR/CD206 protein Leu19-Lys1383 (Thr399Ala) & (Leu407Phe),with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction