missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MMP 8 Protein

Product Code. 15951289
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15951289

Brand: enQuireBio™ QP127065ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the MMP 8 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 4317
Name MMP 8 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol MMP8
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSLTPGNPK WERTNLTYRI RNYTPQLSEA EVERAIKDAF ELWSVASPLI FTRISQGEAD INIAFYQRDH GDNSPFDGPN GILAHAFQPG QGIGGDAHFD AEETWTNTSA NYNLFLVAAH EFGHSLGLAH SSDPGALMYP NYAFRETSNY SLPQDDIDGI QAIYGLSSNP IQPTGPSTPK PCDPSLTFDA ITTLRGEILF FKDRYFWRRH PQLQRVEMNF ISLFWPSLPT GIQAAYEDFD RDLIFLFKGN QYWALSGYDI LQGYPKDISN YGFPSSVQAI DAAVFYRSKT YFFVNDQFWR YDNQRQFMEP GYPKSISGAF PGIESKVDAV FQQEHFFHVF SGPRYYAFDL IAQRVTRVAR GNKWLNCRYG
Buffer The MMP 8 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.