missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human MMP-10 Protein, CF
Beschreibung
Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Catalog # ES002 ). The specific activity is >190 pmol/min/μg, as measured under the described conditions.
Spezifikation
Spezifikation
| Accession Number | P09238 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in MES, NaCl, CaCl2 and Brij-35. |
| Gene ID (Entrez) | 4319 |
| Molecular Weight (g/mol) | M.W. (Observed): 58 kDa and 48 kDa, reducing conditions, M.W. (Theoretical): 52 kDa (pro) & 43 kDa (active) |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived human MMP-10 protein Tyr18-Cys476 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2 |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?