missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human MKK6/MEK6 His Protein

Product Code. 18354254
Change view
Click to view available options
Quantity:
0.01 mg
0.02 mg
0.05 mg
0.1 mg
0.25 mg
0.5 mg
Unit Size:
0.01mg
0.02mg
0.05mg
0.1mg
0.25mg
0.5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354254 0.25 mg 0.25mg
18354773 0.5 mg 0.5mg
18337743 0.1 mg 0.1mg
18345683 0.05 mg 0.05mg
18315362 0.02 mg 0.02mg
18356742 0.01 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 18354254 Supplier Novus Biologicals™ Supplier No. NBP3119630.25MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/ml
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 20% glycerol
Gene ID (Entrez) 5608
Molecular Weight (g/mol) M.W. Theoretical: 38.3 kDa
Quantity 0.25 mg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Gene Alias MAP kinase kinase 6, MAPK/ERK kinase 6, MAPKK6dual specificity mitogen-activated protein kinase kinase 6, MEK6MAPKK 6, mitogen-activated protein kinase kinase 6, MKK6MEK 6, PRKMK6protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6), SAPKK3EC 2.7.12.2
Gene Symbol MAP2K6
Research Category Angiogenesis, Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Protein Kinase
Purity or Quality Grade >90%, by SDS-PAGE
Protein MKK6/MEK6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.