missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MKI67 Protein
A DNA sequence encoding the Homo sapiens (Human) MKI67, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6361-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001139438.1 | |
MKI67 Protein | |
Research Use Only | |
MKI67 | |
Recombinant Protein | |
E. coli | |
NEKKPMKTSP EMDIQNPDDG ARKPIPRDKV TENKRCLRSA RQNESSQPKV AEESGGQKSA KVLMQNQKGK GEAGNSDSMC LRSRKTKSQP AASTLESKSV QRVTRSVKRC AENPKKAEDN VCVKKIRTRS HRDSEDI | |
Greater than 90% as determined by SDS-PAGE. |
4288 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |