missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MKI67 Protein

A DNA sequence encoding the Homo sapiens (Human) MKI67, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6361-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16012750

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001139438.1
MKI67 Protein
Research Use Only
MKI67
Recombinant Protein
E. coli
NEKKPMKTSP EMDIQNPDDG ARKPIPRDKV TENKRCLRSA RQNESSQPKV AEESGGQKSA KVLMQNQKGK GEAGNSDSMC LRSRKTKSQP AASTLESKSV QRVTRSVKRC AENPKKAEDN VCVKKIRTRS HRDSEDI
Greater than 90% as determined by SDS-PAGE.
4288
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers