missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mitogen-activated protein kinase 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Mitogen-activated protein kinase 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6340-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035145.1 | |
Mitogen-activated protein kinase 3 Protein | |
Research Use Only | |
MAPK3 | |
Recombinant Protein | |
E. coli | |
GGEPRRTEGV GPGVPGEVEM VKGQPFDVGP RYTQLQYIGE GAYGMVSSAY DHVRKTRVAI KKISPFEHQT YCQRTLREIQ ILLRFRHENV IGIRDILRAS TLEAMRDVYI VQDLMETDLY KLLKSQQLSN DHICYFLYQI LRGLKYIHSA NVLHRDLKPS NLLINTTCDL KICDFGLARI ADPEHDHTGF LTEYVATRWY RAPEIMLNSK GYTKSIDIWS VGCILAEMLS NRPIFPGKHY LDQLNHILGI LGSPSQEDLN CIINMKARNY LQSLPSKTKV AWAKLFPKSD SKALDLLDRM LTFNPNKRIT VEEALAHPYL EQYYDPTDEP VAEEPFTFAM ELDDLPKERL KELIFQETAR FQPGVLEAP | |
Greater than 90% as determined by SDS-PAGE. |
5595 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |