missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MIP-3 beta / CCL19 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5258-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985779

  • £191.00 / 20µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006265.1
MIP-3 beta / CCL19 Protein
Research Use Only
CCL19
Recombinant Protein
E. coli
GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS
Greater than 95% as determined by SDS-PAGE.
6363
20 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Human MIP-3 beta / CCL19 induces Primary human T cell chemotaxis.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers