missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MIF / GLIF Protein

Product Code. 15925829
Click to view available options
Quantity:
1 mg
25 μg
5 μg
500 μg
Unit Size:
1mg
25µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 15925829

Brand: enQuireBio™ QP52765ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_002406.1
Gene ID (Entrez) 4282
Name MIF / GLIF Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol MIF
Biological Activity Human MIF / GLIF induces IL-8 production in human PBMCs. Activity was further confirmed by measuring chemotaxisof U937 cells via agarose microdroplet assay.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MPMFIVNTNV PRASVPDGFL SELTQQLAQA TGKPPQYIAV HVVPDQLMAF GGSSEPCALC SLHSIGKIGG AQNRSYSKLL CGLLAERLRI SPDRVYINYY DMNAANVGWN NSTFA
Buffer 10 mM sodium phosphate, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.