missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human MICL/CLEC12A Fc Chimera Protein

Product Code. 18745816
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18745816

Brand: R&D Systems™ 10835ML050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human Integrin alpha X beta 2 Protein (Catalog # 5755-AX ) is immobilized at 5.00 μg/mL, 100 μL/well, the concentration of Recombinant Human MICL/CLEC12A Fc Chimera (Catalog# 10835-ML) that produces 50% of the optimal binding response is approximately 1.20-12.0 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAI26292.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 160364
Molecular Weight (g/mol) M.W.-observed: 70-85 kDa, under reducing conditions, M.W.-theroretical: 50 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human MICL/CLEC12A protein MD Human IgG1 (Pro100-Lys330) IEGR Human MICL/CLEC12A (Thr67-Ala265) (N-terminus) Accession # AAI26292.1 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein MICL/CLEC12A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.