missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human MICA His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
MICA Protein is fully biologically active when compared to standard. The specific activity is determined by binding MICA antibody in ELISA.
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | 0.2 um filtered concentrated sterile solution in PBS, pH 7.4, and 8 M Urea. |
| Gene ID (Entrez) | 100507436 |
| Molecular Weight (g/mol) | 36.9 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of MICA as determined by LAL method. |
| Gene Alias | FLJ60820, MGC111087, PERB11.1 |
| Gene Symbol | MICA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction