missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MICA Protein is fully biologically active when compared to standard. The specific activity is determined by binding MICA antibody in ELISA.
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | 0.2 um filtered concentrated sterile solution in PBS, pH 7.4, and 8 M Urea. |
| Gene ID (Entrez) | 100507436 |
| Molecular Weight (g/mol) | 36.9 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of MICA as determined by LAL method. |
| Gene Alias | FLJ60820, MGC111087, PERB11.1 |
| Gene Symbol | MICA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?