missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human MIA Protein
Click to view available options
:
Recombinant Human MIA Protein, Carrier-Free
Description
The Recombinant Human MIA Protein is derived from E. coli. The Recombinant Human MIA Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Source | E. coli-derived human MIA protein Gly25-Gln131,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction