missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human mGluR3 Protein

Product Code. 18726259
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18726259

Brand: R&D Systems™ 9527UR100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Recombinant HumanmgluR3 Protein CF are coupled to effector systems through GTP-binding proteins and modulate glutamate neurotransmission in the central and peripheral nervous systems.

The Recombinant Human mGluR3 Protein is derived from CHO.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14832-1
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 2913
Molecular Weight (g/mol) 56kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line,CHO-derived human mGluR3 protein Asp25-Ser507,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias GLUR3, glutamate receptor, metabotropic 3, GPRC1C, GPRC1Cglutamate metabotropic receptor 3, GRM3,mglu3,mgluR3,mgLUR3metabotropic glutamate receptor 3
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.