missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MFAP4 Protein

A cDNA sequence encoding the MFAP4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12691-HIS-EC-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991239

  • £2975.00 / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MFAP4 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMVSGIR GDALERFCLQ QPLDCDDIYA QGYQSDGVYL IYPSGPSVPV PVFCDMTTEG GKWTVFQKRF NGSVSFFRGW NDYKLGFGRA DGEYWLGLQN MHLLTLKQKY ELRVDLEDFE NNTAYAKYAD FSISPNAVSA EEDGYTLFVA GFEDGGAGDS LSYHSGQKFS TFDRDQDLFV QNCAALSSGA FWFRSCHFAN LNGFYLGGSH LSYANGINWA QWKGFYYSLK RTEMKIRRA
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MFAP4 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers