missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human MFAP4 Protein

Product Code. 18718586
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18718586

Brand: R&D Systems™ 10230MF050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Human MFAP4 is immobilized at 0.5 μg/mL (100 μL/well), the concentration of Recombinant Human Fibrillin-1/FBN1 Fc Chimera that produces 50% of the optimal binding response is 0.1 to 0.6 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P55083
For Use With (Application) Bioactivity
Gene ID (Entrez) 4239
Molecular Weight (g/mol) 28 kDa
Name MFAP-4, microfibril-associated glycoprotein 4, microfibrillar-associated protein 4
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human MFAP4 protein Val22-Ala255, with an N-terminal c-Myc Tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.