missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Methylenetetrahydrofolate reductase Protein
A DNA sequence encoding the Homo sapiens (Human) Methylenetetrahydrofolate reductase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6391-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001317287.1 | |
Methylenetetrahydrofolate reductase Protein | |
Research Use Only | |
MTHFR | |
Recombinant Protein | |
E. coli | |
MVNEARGNSS LNPCLEGSAS SGSESSKDSS RCSTPGLDPE RHERLREKMR RRLESGDKWF SLEFFPPRTA EGAVNLISRF DRMAAGGPLY IDVTWHPAGD PGSDKETSSM MIASTAVNYC GLETILHMTC CRQRLEEITG HLHKAKQLGL KNIMALRGDP IGDQWEEEEG GFNYAVDLVK HIRSEFGDYF DICVAGYPKG HPEAGSFEAD LKHLKEKVSA GADFIITQLF FEADTFFRFV KACTDMGITC PIVPGIFPIQ GYHSLRQLVK LSKLEVPQEI KDVIEPIKDN DAAIRNYGIE LAVSLCQELL ASGLVPGLHF YTLNREMATT EVLKRLGMWT EDPRRPLPWA LSAHPKRREE DVRPIFWASR PKSYIYRTQE WDEFPNGRWG NSSSPAFGEL KDYYLFYLKS KSPKEELLKM WGEELTSEES VFEVFVLYLS GEPNRNGHKV TCLPWNDEPL AAETSLLKEE LLRVNRQGIL TINSQPNING KPSSDPIVGW GPSGGYVFQK AYLEFFTSRE TAEALLQVLK KYELRVNYHL VNVKGENITN APELQPNAVT WGIFPGREII QPTVVDPVSF MFWKDEAFAL WIERWGKLYE EESPSRTIIQ YIHDNYFLVN LVDNDFPLDN CLWQVVEDTL ELLNRPTQNA RETEAP | |
Greater than 90% as determined by SDS-PAGE. |
4524 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |