missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Methionine Aminopeptidase 1/METAP1 Protein
Shop All Bio Techne ProductsDescription
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-386 of Human Methionine Aminopeptidase 1/METAP1 The Recombinant Human Methionine Aminopeptidase 1/METAP1 Protein is derived from E. coli. The Recombinant Human Methionine Aminopeptidase 1/METAP1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1 mg/mL |
| Formulation | 20 mM Tris-HCl buffer (pH 8.0) containing 3 mM DTT, 40% glycerol, 200 mM NaCl, 0.1 mM PMSF |
| Gene ID (Entrez) | 23173 |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4° short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) |
| Regulatory Status | RUO - research use only |
| Species | Human |
| Recombinant | Recombinant |
| Purity or Quality Grade | >90%, by SDS-PAGE |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?