missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MESDC2 Protein
A cDNA sequence encoding the MESDC2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | MESDC2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971209
|
enQuireBio™
QP12682-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951209
|
enQuireBio™
QP12682-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961209
|
enQuireBio™
QP12682-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MESDC2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MESDC2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAEGSPGTPD ESTPPPRKKK KDIRDYNDAD MARLLEQWEK DDDIEEGDLP EHKRPSAPVD FSKIDPSKPE SILKMTKKGK TLMMFVTVSG SPTEKETEEI TSLWQGSLFN ANYDVQRFIV GSDRAIFMLR DGSYAWEIKD FLVGQDRCAD VTLEGQVYPG KGGGSKEKNK TKQDKGKKKK EGDLKSRSSK EENRAGNKRE DL | |
Greater than 90.0% as determined by SDS-PAGE. |