missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Malic Enzyme activity was assayed spectrophotometrically at 340nm as described in Mandela and Sauer (1975). The standard reaction mixture contained 50 mM Tris-HCl, 3 mM MnCl2, 5 mM malate, 0.12 mM NADP+, 2.5 mM fumarate. Assay was performed in a Beckman spectrophotometer. The Km value is 1.5 +/- 0.6 mM.
Specifications
Specifications
| Accession Number | P23368 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 4200 |
| Molecular Weight (g/mol) | 64.4 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of ME2 as determined by LAL method. |
| Gene Alias | EC 1.1.1, EC 1.1.1.38, malate dehydrogenase, Malic enzyme 2, malic enzyme 2, NAD(+)-dependent, mitochondrial, NAD-dependent malic enzyme, mitochondrial, NAD-ME, ODS1, pyruvic-malic carboxylase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?