missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MCEE Protein
A cDNA sequence encoding the MCEE was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12659-5ug
Additional Details : Weight : 0.01000kg
Specifications
84693 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQVTGSVWNL GRLNHVAIAV PDLEKAAAFY KNILGAQVSE AVPLPEHGVS VVFVNLGNTK MELLHPLGRD SPIAGFLQKN KAGGMHHICI EVDNINAAVM DLKKKKIRSL SEEVKIGAHG KPVIFLHPKD CGGVLVELEQ A | |
Greater than 90% as determined by SDS-PAGE. |
MCEE Protein | |
Research Use Only | |
MCEE | |
Human | |
His | |
The MCEE protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 1mM DTT, 0.1mM PMSF and 10% glycerol. |