missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAX Protein
A cDNA sequence encoding the MAX was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12653-25ug
Additional Details : Weight : 0.01000kg
Specifications
MAX Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSDNDDIEVE SDEEQPRFQS AADKRAHHNA LERKRRDHIK DSFHSLRDSV PSLQGEKASR AQILDKATEY IQYMRRKNHT HQQDIDDLKR QNALLEQQVR ALEKARSSAQ LQTNYPSSDN SLYTNAKGST ISAFDGGSDS SSESEPEEPQ SRKKLRMEAS LEHHHHHH | |
Greater than 90% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAX Human solution containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |