missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MARCKS-related Protein

A DNA sequence encoding the Homo sapiens (Human) MARCKS-related protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6346-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16052640

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_075385.1
MARCKS-related Protein
Research Use Only
MARCKSL1
Recombinant Protein
E. coli
MGSQSSKAPR GDVTAEEAAG ASPAKANGQE NGHVKSNGDL SPKGEGESPP VNGTDEAAGA TGDAIEPAPT SQGAEAKGEV PPKETPKKKK KFSFKKPFKL SGLSFKRNRK EGGGDSSASS PTEEEQEQGE IGACSDEGTA QEGKAAATPE SQEPQAKGAE ASAASEEEAG PQATEPSTPS GPESGPTPAS AEQNE
Greater than 80% as determined by SDS-PAGE.
65108
50 μg
Not determined.
Full Length of BC066915
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers