missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAPRE3 Protein
A cDNA sequence encoding the MAPRE3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12646-1mg
Additional Details : Weight : 0.01000kg
Specifications
MAPRE3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAVNVYSTSV TSENLSRHDM LAWVNDSLHL NYTKIEQLCS GAAYCQFMDM LFPGCVHLRK VKFQAKLEHE YIHNFKVLQA AFKKMGVDKI IPVEKLVKGK FQDNFEFIQW FKKFFDANYD GKDYNPLLAR QGQDVAPPPN PGDQIFNKSK KLIGTAVPQR TSPTGPKNMQ TSGRLSNVAP PCILRKNPPS ARNGGHETDA QILELNQQLV DLKLTVDGLE KERDFYFSKL RDIELICQEH ESENSPVISG IIGILYATEE GFAPPEDDEI EEHQQEDQDE Y | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MAPRE3 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 2mM DTT, 0.1M NaCl and 10% glycerol. |