missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mannose-6-phosphate isomerase Protein

A DNA sequence encoding the Homo sapiens (Human) Mannose-6-phosphate isomerase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6375-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002850

  • £384.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001276085.1
Mannose-6-phosphate isomerase Protein
Research Use Only
MPI
Recombinant Protein
E. coli
AAPRVFPLSC AVQQYAWGKM GSNSEVARLL ASSDPLAQIA EDKPYAELWM GTHPRGDAKI LDNRISQKTL SQWIAENQDS LGSKVKDTFN GNLPFLFKVL SVETPLSIQA HPNKELAEKL HLQAPQHYPD ANHKPEMAIA LTPFQGLCGF RPVEEIVTFL KKVPEFQFLI GDEAATHLKQ TMSHDSMS-Q AVASSLQSCF SHLMKSEKKV VVEQLNLLVK RISQQAAAGN NMEDIFGELL LQLHQQYPGD IGCFAIYFLN LLTLKPGEAM FLEANVPHAY LKGDCVECMA CSDNTVRAGL TPKFIDVPTL CEMLSYTPSS SKDRLFLPTR SQEDPYLSIY DPPVPDFTIM KTEVPGSVTE YKVLALDSAS ILLMVQGTVI ASTPTTQTPI PLQRGGVLFI GANESVSLKL TEPKDLLIFR ACCLL
Greater than 80% as determined by SDS-PAGE.
4351
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers