missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAGEB10 Protein
A cDNA sequence encoding the MAGEB10 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 139422 |
---|---|
Name | MAGEB10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | MAGEB10 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951039
|
enQuireBio™
QP12627-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941039
|
enQuireBio™
QP12627-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931039
|
enQuireBio™
QP12627-100UG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
139422 | |
Research Use Only | |
MAGEB10 | |
Human | |
His | |
MAGEB10 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol, 1mM DTT and 0.1mM PMSF. |
MAGEB10 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPRGQKS KLRAREKRRQ ARGGLEDLID ALDILEEEEE SPPSASACLK DVFQSSLDGA SNNPHGLREA QSTSTSATAA SHTRHPEGVN DQMEERPICT QDLEATDSFP RGPVDEKVII LVHYLLYKYQ MKEPITKADM LRNVTQMSKS QFPVILSRAS EHLELIFGLD LKEVEPNKHI YVLVNKLDLG CDAKLSDETG VPKTGLLMTV LGIIFTNGNC VAEEEVWKVF NTMGLYDGIE HFMFGEPRKL LTKDLVKENY LEYQQVPNSD PPRYQFLWGP RAHAETSKMK VLEFLAKVND TAPSEFSNWY TEALQDEEER ARARVAAKAR VSATAGARSK VKSSKSSQLQ | |
Greater than 85% as determined by SDS-PAGE. |