missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human MAG/Siglec-4a Fc Chimera Protein

Product Code. 18737716
Click to view available options
:
50μg
This item is not returnable. View return policy

Product Code. 18737716

Brand: R&D Systems™ 8940MG050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human MAG/Siglec-4a Fc Chimera Protein is derived from HEK293. The Recombinant Human MAG/Siglec-4a Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 4099
Molecular Weight (g/mol) Theoretical M.W.: 81kDa
Quantity 50μg
Source Human embryonic kidney cell,HEK293-derived human MAG/Siglec-4a protein, (N-terminus) Human MAG (Gly20-Pro516) Accession &Num; P20916 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias GMAsialic acid binding Ig-like lectin 4A, myelin associated glycoprotein, myelin-associated glycoprotein, sialic acid-binding immunoglobulin-like lectin 4A, Siglec4a, SIGLEC-4A, S-MAG
Common Name MAG/Siglec-4a
Gene Symbol MAG
Biological Activity Measured by its ability to inhibit neurite outgrowth of dissociated E13 chick embryonic dorsal root ganglia (DRG) neurons. When immobilized as a 3μL droplet containing 200 ng on a nitrocellulose coated microplate, Recombinant Human MAG/Siglec-4a Fc Chimera is able to significantly inhibit neurite outgrowth.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human MAG (Gly20-Pro516) Accession # P20916, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.