missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAF1 Protein
A DNA sequence encoding the Homo sapiens (Human) MAF1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8112-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_115648.2 | |
MAF1 Protein | |
Research Use Only | |
MAF1 | |
Recombinant Protein | |
E. coli | |
MKLLENSSFE AINSQLTVET GDAHIIGRIE SYSCKMAGDD KHMFKQFCQE GQPHVLEALS PPQTSGLSPS RLSKSQGGEE EGPLSDKCSR KTLFYLIATL NESFRPDYDF STARSHEFSR EPSLSWVVNA VNCSLFSAVR EDFKDLKPQL WNAVDEEICL AECDIYSYNP DLDSDPFGED GSLWSFNYFF YNKRLKRIVF FSCRSISGST YTPSEAGNEL DMELGEEEVE EESRSGGSGA EETSTMEEDR VPVICI | |
Greater than 90% as determined by SDS-PAGE. |
84232 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |