missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human M-CSF Protein

Product Code. 18146970
Click to view available options
Quantity:
1 mg
25 μg
5 μg
500 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
1mg
25µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18146970

Brand: R&D Systems™ 216MC025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using M-NFS-60 mouse myelogenous leukemia lymphoblast cells. Nakoinz, I. et al. (1990) J. Immunol. 145:860. The ED50 for this effect is 0.5-1.5 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 18.5 kDa (monomer)
Name M-CSF
Quantity 25 μg
Source E. coli-derived human M-CSF protein Glu33-Ser190,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 50-500μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Purity or Quality Grade 97%, by SDS-PAGE with silver staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.