missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Especificaciones
Especificaciones
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 116068 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 40.7 kDa |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Avoid freeze/thaw cycles. |
| Gene Alias | LysM, putative peptidoglycan-binding, domain containing 3 |
| Gene Symbol | LYSMD3 |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | LYSMD3 |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?