missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lys-63-specific deubiquitinase BRCC36 Protein
A DNA sequence encoding the Homo sapiens (Human) Lys-63-specific deubiquitinase BRCC36, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5727-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001018065.1 | |
Lys-63-specific deubiquitinase BRCC36 Protein | |
Research Use Only | |
BRCC3 | |
Recombinant Protein | |
E. coli | |
AVQVVQAVQA VHLESDAFLV CLNHALSTEK EEVMGLCIGE LNDDTRSDSK FAYTGTEMRT VAEKVDAVRI VHIHSVIILR RSDKRKDRVE ISPEQLSAAS TEAERLAELT GRPMRVVGWY HSHPHITVWP SHVDVRTQAM YQMMDQGFVG LIFSCFIEDK NTKTGRVLYT CFQSIQAQKS SESLHGPRDF WSSSQHISIE GQKEEERYER IEIPIHIVPH VTIGKVCLES AVELPKILCQ EEQDAYRRIH SLTHLDSVTK IHNGSVFTKN LCSQMSAVSG PLLQWLEDRL EQNQQHLQEL QQEKEELMQE LSSLE | |
Greater than 90% as determined by SDS-PAGE. |
79184 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |