missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Lymphotoxin alpha1/beta2 Protein

Product Code. 18797907
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18797907

Brand: R&D Systems™ 8884LY025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Lymphotoxin alpha1/beta2 Protein is derived from HEK293. The Recombinant Human Lymphotoxin alpha1/beta2 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 4049
Molecular Weight (g/mol) Observed M.W.: 65-76kDa, reducing conditions, Theoretical M.W.: 61kDa
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived human Lymphotoxin protein, (N-terminus) rhLT alpha (Leu35-Leu205) Accession &Num; P01374 GGGGS rhLT beta (Gln49-Gly244) Accession &Num; Q06643 GGGGS rhLT beta (Gln49-Gly244) Accession &Num; Q06643 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name Lymphotoxin
Gene Symbol LTA
Biological Activity Measured by its ability to induce IL-8 secretion by A375 human melanoma cells. The ED50 for this effect is typically 3-15ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived rhLT alpha (Leu35-Leu205) Accession # P01374, N-terminus; GGGGS; rhLT beta (Gln49-Gly244) Accession # Q06643; GGGGS; rhLT beta (Gln49-Gly244) Accession # Q06643, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.