missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LY86 Protein

Product Code. 15940959
Change view
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15940959 1 mg 1mg
15960959 5 μg 5µg
15950959 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15940959 Supplier enQuireBio™ Supplier No. QP126021mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the LY86 was constructed and used to recombinantly synthesize the protein.

Specifications

Name LY86 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSGGGGKAW PTHVVCSDSG LEVLYQSCDP LQDFGFSVEK CSKQLKSNIN IRFGIILRED IKELFLDLAL MSQGSSVLNF SYPICEAALP KFSFCGRRKG EQIYYAGPVN NPEFTIPQGE YQVLLELYTE KRSTVACANA TIMCS
Buffer The LY86 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.