missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lupus La Protein
A DNA sequence encoding the Homo sapiens (Human) Lupus La protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6731-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001281074.1 | |
Lupus La Protein | |
Research Use Only | |
SSB | |
Recombinant Protein | |
E. coli | |
MAENGDNEKM AALEAKICHQ IEYYFGDFNL PRDKFLKEQI KLDEGWVPLE IMIKFNRLNR LTTDFNVIVE ALSKSKAELM EISEDKTKIR RSPSKPLPEV TDEYKNDVKN RSVYIKGFPT DATLDDIKEW LEDKGQVLNI QMRRTLHKAF KGSIFVVFDS IESAKKFVET PGQKYKETDL LILFKDDYFA KKNEERKQNK VEAKLRAKQE QEAKQKLEED AEMKSLEEKI GCLLKFSGDL DDQTCREDLH ILFSNHGEIK WIDFVRGAKE GIILFKEKAK EALGKAKDAN NGNLQLRNKE VTWEVLEGEV EKEALKKIIE DQQESLNKWK SKGRRFKGKG KGNKAAQPGS GKGKVQFQGK KTKFASDDEH DEHDENGATG PVKRAREETD KEEPASKQQK TENGAGDQ | |
Greater than 90% as determined by SDS-PAGE. |
6741 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |