missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LTF Holo Protein

Product Code. p-7150468
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
This item is not returnable. View return policy

Product Code. 15950939

Brand: enQuireBio™ QP1259610mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the LTF Holo was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 4057
Name LTF Holo Protein
Quantity 10 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol LTF
Product Type Recombinant Protein
Cross Reactivity Human
Species Rice
Protein Tag Untagged
Sequence MKLVFLVLLF LGALGLCLAG RRRSVQWCAV SQPEATKCFQ WQRNMRKVRG PPVSCIKRDS PIQCIQAIAE NRADAVTLDG GFIYEAGLAP YKLRPVAAEV YGTERQPRTH YYAVAVVKKG GSFQLNELQG LKSCHTGLRR TAGWNVPIGT LRPFLNWTGP PEPIEAAVAR FFSASCVPGA DKGQFPNLCR LCAGTGENKC AFSSQEPYFS YSGAFKCLRD GAGDVAFIRE STVFEDLSDE AERDEYELLC PDNTRKPVDK FKDCHLARVP SHAVVARSVN GKEDAIWNLL RQAQEKFGKD KSPKFQLFGS PSGQKDLLFK DSAIGFSRVP PRIDSGLYLG SGYFTAIQNL RKSEEEVAAR RARVVWCAVG EQELRKCNQW SGLSEGSVTC SSASTTEDCI ALVLKGEADA MSLDGGYVYT AGKCGLVPVL AENYKSQQSS DPDPNCVDRP VEGYLAVAVV RRSDTSLTWN SVKGKKSCHT AVDRTAGWNI PMGLLFNQTG SCKFDEYFSQ SCAPGSDPRS NLCALCIGDE QGENKCVPNS NERYYGYTGA FRCLAENAGD VAFVKDVTVL QNTDGNNNEA WAKDLKLADF ALLCLDGKRK PVTEARSCHL AMAPNHAVVS RMDKVERLKQ VLLHQQAKFG RNGSDCPDKF CLFQSETKNL LFNDNTECLA RLHGKTTYEK YLGPQYVAGI TNLKKCSTSP LLEACEFLRK
Buffer The Human Holo lactotransferrin was lyophilized from water, contains 1.66 mg iron/gram Lactoferrin.
Purity or Quality Grade Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.