missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LTA Protein

Product Code. 15914969
Click to view available options
Quantity:
10 μg
100 μg
250 μg
50 μg
Unit Size:
100µg
10µg
250µg
50µg
This item is not returnable. View return policy

Product Code. 15914969

Brand: enQuireBio™ QP1387ec50ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Human LTA, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000586.2
Gene ID (Entrez) 4049
Name LTA Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol LTA
Protein Length Partial (see sequence information for more details).
Biological Activity The ED50 as determined by a cytotoxicity assay using murine L929 cells is less than 5 pg/ml, corresponding to a specific activity of > 2.0 M 108 IU/mg in the presence of actinomycin D.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence LPGVGLTPSA AQTARQHPKM HLAHSTLKPA AHLIGDPSKQ NSLLWRANTD RAFLQDGFSL SNNSLLVPTS GIYFVYSQVV FSGKAYSPKA TSSPLYLAHE VQLFSSQYPF HVPLLSSQKM VYPGLQEPWL HSMYHGAAFQ LTQGDQLSTH TDGIPHLVLS PSTVFFGAFA L
Buffer 0.2 um filtered PBS, pH7.4, with 0.02 % Tween-20, lyophilized
Purity or Quality Grade Greater than 96% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.