missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human LT beta R/TNFRSF3 Fc Chimera (CHO)
Description
The Recombinant Human LT beta R/TNFRSF3 Fc Chimera (CHO) is derived from CHO. The Recombinant Human LT beta R/TNFRSF3 Fc Chimera (CHO) has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 4055 |
| Molecular Weight (g/mol) | 48.5 kDa (monomer) |
| Name | Lymphotoxin beta R/TNFRSF3 Fc Chimera |
| Quantity | 100μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Lymphotoxin beta R/TNFRSF3 protein, (N-terminus) Human Lymphotoxin beta R (Ser28-Met227) Accession &Num; NP_002333 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?