missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LRRC59 Protein
A cDNA sequence encoding the LRRC59 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12587-10ug
Additional Details : Weight : 0.01000kg
Specifications
LRRC59 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTKAGSK GGNLRDKLDG NELDLSLSDL NEVPVKELAA LPKATILDLS CNKLTTLPSD FCGLTHLVKL DLSKNKLQQL PADFGRLVNL QHLDLLNNKL VTLPVSFAQL KNLKWLDLKD NPLDPVLAKV AGDCLDEKQC KQCANKVLQH MKAVQADQER ERQRRLEVER EAEKKREAKQ RAKEAQEREL RKREKAEEKE RRRKEYDALK AAKREQEKKP KKEANQAPKS KSGSRPRKPP PRKHTRS | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LRRC59 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT, 2mM EDTA and 50% glycerol. |