missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human LOX-1/OLR1 Protein

Product Code. 18119379 Shop All Bio Techne Products
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
This item is not returnable. View return policy

Product Code. 18119379

Brand: Novus Biologicals™ NBP25307450UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 58-273 of Human LOX-1/OLR1 The Recombinant Human LOX-1/OLR1 Protein is derived from Baculovirus. The Recombinant Human LOX-1/OLR1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 4973
Molecular Weight (g/mol) 25.8kDa
Name Human LOX-1/OLR1 Protein
Purification Method >95%, by PAGE
Quantity 50 μg
Source Baculovirus
Immunogen ADPMQLSQVS DLLTQEQANL THQKKKLEGQ ISARQQAEEA SQESENELKE MIETLARKLN EKSKEQMELH HQNLNLQETL KRVANCSAPC PQDWIWHGEN CYLFSSGSFN WEKSQEKCLS LDAKLLKINS TADLDFIQQA ISYSSFPFWM GLSRRNPSYP WLWEDGSPLM PHLFRVRGAV SQTYPSGTCA YIQRGAVYAE NCILAAFSIC QKKANLRAQH HHHHH
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX-1, LOX1ox LDL receptor 1, LOXIN, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1
Gene Symbol OLR1
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.