missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Lipocalin-2/NGAL Protein
Description
The Recombinant Human Lipocalin-2/NGAL Protein is derived from NS0. The Recombinant Human Lipocalin-2/NGAL Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Accession Number | P80188 |
| Formulation | Supplied as a 0.2μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 3934 |
| Molecular Weight (g/mol) | 22 kDa |
| Name | Lipocalin-2/NGAL |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Lipocalin-2/NGAL protein Gln21-Gly198,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Gene Alias | 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?