missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LIN7B Protein

A cDNA sequence encoding the LIN7B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12574-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960859

  • £155.00 / 10µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

LIN7B Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAALVEP LGLERDVSRA VELLERLQRS GELPPQKLQA LQRVLQSRFC SAIREVYEQL YDTLDITGSA EIRAHATAKA TVAAFTASEG HAHPRVVELP KTDEGLGFNI MGGKEQNSPI YISRVIPGGV ADRHGGLKRG DQLLSVNGVS VEGEQHEKAV ELLKAAQGSV KLVVRYTPRV LEEMEARFEK MRSARRRQQH QSYSSLESRG
Greater than 90.0% as determined by SDS-PAGE
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
LIN7B protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers