missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human LILRB3/CD85a/ILT5 Fc Chimera Protein

Product Code. 18167870
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18167870

Brand: R&D Systems™ 1806T5050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to bind fluorescein-conjugated S. aureus Bioparticles. Jiang, Y. et al. (2006) J. Biol. Chem. 281:11834. The ED50 for this effect is 0.4-2 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAB68668
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 11025
Molecular Weight (g/mol) M.W. (Observed): 95-105 kDa, reducing conditions; M.W. (theoretical): 72.6 kDa (monomer)
Name ILT5/CD85a Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human LILRB3/CD85a/ILT5 protein Human ILRB3/CD85a/ILT5 (Gly24-Glu443) (N-terminus) Accession # AAB68668 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol LILRB3
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein ILT5/CD85a/LILRB3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.