missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human LILRB2/CD85d/ILT4 Fc Chimera Protein

Product Code. 18379437
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379437 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379437 Supplier R&D Systems™ Supplier No. 2078T4050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human Angiopoietin-like Protein 5/ANGPTL5 (Catalog # 6675-AN ) is coated at 4 μg/mL, Recombinant Human LILRB2/CD85d/ILT4 Fc Chimera binds with a typical ED50 of 0.15-0.9 μg/mL. Measured by its ability to support the adhesion of HSB2 human peripheral blood acute lymphoblastic leukemia cells. Recombinant Human LILRB2/CD85d/ILT4 Fc Chimera immobilized at 35 μg/mL, 100 μL/well, will mediate >40% HSB-2 cell adhesion (105 cells/well).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N423
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 10288
Molecular Weight (g/mol) M.W. (Observed): 94-100 kDa, reducing conditions; M.W. (theoretical): 73.9 kDa (monomer)
Name ILT4/CD85d Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human LILRB2/CD85d/ILT4 protein Human LILRB2/CD85d/ILT4 (Gly24-His458) Accession # (N-terminus)AAH36827.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2 subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10
Gene Symbol LILRB2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ILT4/CD85d/LILRB2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.